CH4183975746 ChemBridge Corporation

5-bromo-1H-1,2,4-triazole

  • MDL number MFCD09055371
  • Cas Number : 7343-33-1
  • Molecular Weight : 147.9614

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4050415

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4050415
Availability details for
Quantity
  • Availability
  • Add to Cart