CH4196934255 ChemBridge Corporation

3-bromo-5-methyl-2-(4H-1,2,4-triazol-4-yl)pyridine

  • MDL number MFCD12198490
  • Cas Number : 1123169-57-2
  • Molecular Weight : 239.072

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data FREE
Assay 0.95
Storage temp. Room Temperature

Safety Information

msds https://www.hit2lead.com/building-blocks/4034334
Availability details for
Quantity
  • Availability
  • Add to Cart