CH4196934270 ChemBridge Corporation

(1E)-1-(4-bromophenyl)-3-(4-methylphenyl)triaz-1-ene

  • MDL number MFCD00638810
  • Cas Number : 22715-74-8
  • Molecular Weight : 290.1585

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5511892

Safety Information

MSDS https://www.hit2lead.com/sds/5511892_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart