CH4196934271 ChemBridge Corporation

2-chloro-4-(4H-1,2,4-triazol-4-yl)benzoic acid

  • MDL number MFCD06809616
  • Cas Number : 842977-29-1
  • Molecular Weight : 223.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4023533

Safety Information

MSDS https://www.hit2lead.com/sds/4023533_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart