CH4196934282 ChemBridge Corporation

1-methyl-1H-pyrazole-4-carbonitrile

  • MDL number MFCD04969704
  • Cas Number : 66121-71-9
  • Molecular Weight : 107.1133

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4027649

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/sds/4027649_sds.pdf
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart