CH4196934291 ChemBridge Corporation

4-(difluoromethoxy)-2-methylaniline

  • MDL number MFCD03422548
  • Cas Number : 39211-57-9
  • Molecular Weight : 173.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4028886

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4028886
Availability details for
Quantity
  • Availability
  • Add to Cart