CH4196934327 ChemBridge Corporation

3-[2-(dimethylamino)ethoxy]benzaldehyde

  • MDL number MFCD09675043
  • Cas Number : 81068-25-9
  • Molecular Weight : 193.2423

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4003879

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4003879
Availability details for
Quantity
  • Availability
  • Add to Cart