CH4196934335 ChemBridge Corporation

5-chloro-6-methyl-1,3-benzothiazol-2-amine

  • MDL number MFCD03725038
  • Cas Number : 50850-98-1
  • Molecular Weight : 198.6726

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4029976

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4029976
Availability details for
Quantity
  • Availability
  • Add to Cart