CH4196934341 ChemBridge Corporation

3-bromo-4-methyl-2-quinolinol

  • MDL number MFCD02315177
  • Cas Number : 37778-22-6
  • Molecular Weight : 238.0806

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data FREE
Assay 0.95
Storage temp. Room Temperature

Safety Information

msds https://www.hit2lead.com/building-blocks/4030122
Availability details for
Quantity
  • Availability
  • Add to Cart