CH4196934379 ChemBridge Corporation

3-(1H-1,2,4-triazol-1-yl)aniline

  • MDL number MFCD08271899
  • Cas Number : 176032-78-3
  • Molecular Weight : 160.1759

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://www.hit2lead.com/building-blocks/4030979

Safety Information

msds https://www.hit2lead.com/building-blocks/4030979
Availability details for
Quantity
  • Availability
  • Add to Cart