CH4196934416 ChemBridge Corporation

4-bromo-2-hydrazino-1,3-benzothiazole

  • MDL number MFCD04448799
  • Cas Number : 872696-03-2
  • Molecular Weight : 244.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://www.hit2lead.com/building-blocks/4031600

Safety Information

msds https://www.hit2lead.com/building-blocks/4031600
Availability details for
Quantity
  • Availability
  • Add to Cart