CH4196934443 ChemBridge Corporation

3-(4-aminophenyl)-N,N-dimethylpropanamide

  • MDL number MFCD10022146
  • Cas Number : 1018284-46-2
  • Molecular Weight : 192.2575

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4032098

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4032098
Availability details for
Quantity
  • Availability
  • Add to Cart