CH4196934480 ChemBridge Corporation

1-(1-propyl-1H-pyrazol-4-yl)methanamine

  • MDL number MFCD04970262
  • Cas Number : 1006333-36-3
  • Molecular Weight : 139.1982

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4032500

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4032500
Availability details for
Quantity
  • Availability
  • Add to Cart