CH4196934484 ChemBridge Corporation

N-methyl-2-(4-piperidinyl)acetamide

  • MDL number MFCD09933326
  • Cas Number : 70724-80-0
  • Molecular Weight : 156.2254

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4032563

Safety Information

MSDS https://www.hit2lead.com/sds/4032563_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart