CH4196934526 ChemBridge Corporation

1-(2-pyrrolidinylmethyl)-1H-pyrazole

  • MDL number MFCD09859232
  • Cas Number : 1171334-97-6
  • Molecular Weight : 151.2089

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4014497

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4014497
Availability details for
Quantity
  • Availability
  • Add to Cart