CH4196934539 ChemBridge Corporation

2-(chloromethyl)-1,4-dioxane

  • MDL number MFCD01994526
  • Cas Number : 21048-16-8
  • Molecular Weight : 136.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(chloromethyl)-1,4-dioxane
Inchi Code 1S/C5H9ClO2/c6-3-5-4-7-1-2-8-5/h5H,1-4H2
Inchi Key SFABQWISNPPOOB-UHFFFAOYSA-N

Description

Purity 95%
Salt Data FREE
Storage temp. -20
Physical Form liquid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/4015243

Safety Information

MSDS https://www.hit2lead.com/sds/4015243_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart