CH4196934557 ChemBridge Corporation

1-(6-chloro-3-pyridazinyl)azepane

  • MDL number MFCD01460065
  • Cas Number : 312318-48-2
  • Molecular Weight : 211.6913

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4033373

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4033373
Availability details for
Quantity
  • Availability
  • Add to Cart