CH4196934585 ChemBridge Corporation

5-(3-fluorophenyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09028454
  • Cas Number : 341978-66-3
  • Molecular Weight : 179.1511

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4033660

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4033660
Availability details for
Quantity
  • Availability
  • Add to Cart