CH4196934597 ChemBridge Corporation

4-chloro-6-methyl-2-phenylpyrimidine

  • MDL number MFCD00296931
  • Cas Number : 29509-92-0
  • Molecular Weight : 204.6556

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4001930

Safety Information

MSDS https://www.hit2lead.com/sds/4001930_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart