CH4196934599 ChemBridge Corporation

6-methyl-4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carboxylic acid

  • MDL number MFCD01690160
  • Cas Number : 32092-27-6
  • Molecular Weight : 204.1821

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4033799

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4033799
Availability details for
Quantity
  • Availability
  • Add to Cart