CH4196934601 ChemBridge Corporation

4-(2-methyl-1H-imidazol-1-yl)-2-butanone

  • MDL number MFCD11190953
  • Cas Number : 120216-60-6
  • Molecular Weight : 152.1937

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data FREE
Assay 0.95
Storage temp. Room Temperature

Safety Information

msds https://www.hit2lead.com/building-blocks/4033806
Availability details for
Quantity
  • Availability
  • Add to Cart