CH4196934623 ChemBridge Corporation

1-acetyl-3-piperidinecarbohydrazide

  • MDL number MFCD11643574
  • Cas Number : 1098343-82-8
  • Molecular Weight : 185.2236

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4034162

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4034162
Availability details for
Quantity
  • Availability
  • Add to Cart