CH4196934648 ChemBridge Corporation

2-amino-1,3-benzothiazole-6-carboxamide

  • MDL number MFCD01466259
  • Cas Number : 111962-90-4
  • Molecular Weight : 193.2257

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4034423

Safety Information

MSDS https://www.hit2lead.com/sds/4034423_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart