CH4196934657 ChemBridge Corporation

4-amino-1-methyl-1H-pyrazole-5-carboxamide

  • MDL number MFCD00464001
  • Cas Number : 92534-73-1
  • Molecular Weight : 140.1432

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4034516

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4034516
Availability details for
Quantity
  • Availability
  • Add to Cart