CH4196934658 ChemBridge Corporation

2-(1H-pyrrol-1-yl)pentanoic acid

  • MDL number MFCD12096890
  • Cas Number : 70901-15-4
  • Molecular Weight : 167.205

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data FREE
Assay 0.95
Storage temp. Room Temperature

Safety Information

msds https://www.hit2lead.com/building-blocks/4034533
Availability details for
Quantity
  • Availability
  • Add to Cart