CH4196934672 ChemBridge Corporation

2-methoxy-2-methylpropanoic acid

  • MDL number MFCD07787581
  • Cas Number : 13836-62-9
  • Molecular Weight : 118.1311

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4021500

Safety Information

MSDS https://www.hit2lead.com/sds/4021500_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart