CH4196934674 ChemBridge Corporation

3-(1-pyrrolidinylcarbonyl)aniline

  • MDL number MFCD09045578
  • Cas Number : 160647-74-5
  • Molecular Weight : 190.2417

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4021618

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4021618
Availability details for
Quantity
  • Availability
  • Add to Cart