CH4196934679 ChemBridge Corporation

(3-isopropyl-4,5-dihydro-5-isoxazolyl)methanol

  • MDL number MFCD12028399
  • Cas Number : 1142210-97-6
  • Molecular Weight : 143.1836

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4035834

Safety Information

MSDS https://www.hit2lead.com/sds/4035834_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart