CH4196934680 ChemBridge Corporation

6-(2-hydroxyethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid

  • MDL number MFCD05997895
  • Cas Number : 712319-12-5
  • Molecular Weight : 207.1861

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4035866

Safety Information

MSDS https://www.hit2lead.com/sds/4035866_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart