CH4196934724 ChemBridge Corporation

6-bromo-4-methyl-2(1H)-quinolinone

  • MDL number MFCD02159765
  • Cas Number : 89446-19-5
  • Molecular Weight : 238.0806

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6183322

Safety Information

MSDS https://www.hit2lead.com/sds/6183322_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart