CH4238555663 ChemBridge Corporation

5-isopropyl-3-(morpholin-4-ylcarbonyl)thiophen-2-amine

  • MDL number MFCD03945259
  • Cas Number : 588714-50-5
  • Molecular Weight : 254.3485

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3001547

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3001547
Availability details for
Quantity
  • Availability
  • Add to Cart