CH4238555692 ChemBridge Corporation

2-(bromomethyl)-2-methyl-1,3-dioxolane

  • MDL number MFCD09034161
  • Cas Number : 33278-96-5
  • Molecular Weight : 181.0278

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3001590

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3001590
Availability details for
Quantity
  • Availability
  • Add to Cart