CH4238555918 ChemBridge Corporation

4-iodo-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD00465280
  • Cas Number : 6647-93-4
  • Molecular Weight : 237.9833

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3001905

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3001905
Availability details for
Quantity
  • Availability
  • Add to Cart