CH4238555921 ChemBridge Corporation

(3S)-1-{[3-(aminomethyl)-1,2,4-oxadiazol-5-yl]carbonyl}pyrrolidin-3-ol

  • MDL number MFCD12026798
  • Cas Number : 1186662-39-4
  • Molecular Weight : 212.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3001908

Safety Information

MSDS https://www.hit2lead.com/sds/3001908_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart