CH4238555967 ChemBridge Corporation

7-amino-3,4-dihydroquinolin-2(1H)-one

  • MDL number MFCD06809628
  • Cas Number : 22246-07-7
  • Molecular Weight : 162.1885

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3001973

Safety Information

MSDS https://www.hit2lead.com/sds/3001973_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart