CH4238556005 ChemBridge Corporation

3-propyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD08144099
  • Cas Number : 1007541-75-4
  • Molecular Weight : 154.1665

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3002026

Safety Information

MSDS https://www.hit2lead.com/sds/3002026_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart