CH4238556073 ChemBridge Corporation

4-bromo-1-ethyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD02090837
  • Cas Number : 514800-97-6
  • Molecular Weight : 219.036

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3002127

Safety Information

MSDS https://www.hit2lead.com/sds/3002127_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart