CH4238556107 ChemBridge Corporation

5-(piperidin-1-ylmethyl)-2-furoic acid

  • MDL number MFCD00455269
  • Cas Number : 301353-36-6
  • Molecular Weight : 209.2417

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3002171

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3002171
Availability details for
Quantity
  • Availability
  • Add to Cart