CH4238556218 ChemBridge Corporation

1-(2-methylbenzyl)-1H-1,2,4-triazol-3-amine

  • MDL number MFCD03422586
  • Cas Number : 832740-49-5
  • Molecular Weight : 188.2291

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3002319

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3002319
Availability details for
Quantity
  • Availability
  • Add to Cart