CH4238556358 ChemBridge Corporation

5-cyclopropyl-1,3,4-thiadiazol-2-amine

  • MDL number MFCD00051649
  • Cas Number : 57235-50-4
  • Molecular Weight : 141.1942

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3002513

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3002513
Availability details for
Quantity
  • Availability
  • Add to Cart