CH4238556407 ChemBridge Corporation

butanohydrazide

  • MDL number MFCD00025135
  • Cas Number : 3538-65-6
  • Molecular Weight : 102.135

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3002586

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3002586
Availability details for
Quantity
  • Availability
  • Add to Cart