CH4238556417 ChemBridge Corporation

4-[(2-methyl-1H-imidazol-1-yl)methyl]aniline

  • MDL number MFCD06797794
  • Cas Number : 772311-98-5
  • Molecular Weight : 187.241

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3002600

Safety Information

MSDS https://www.hit2lead.com/sds/3002600_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart