CH4238556421 ChemBridge Corporation

2-hydroxy-2-phenylacetohydrazide

  • MDL number MFCD00038133
  • Cas Number : 2443-66-5
  • Molecular Weight : 166.1772

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3002604

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3002604
Availability details for
Quantity
  • Availability
  • Add to Cart