CH4238556423 ChemBridge Corporation

3-(morpholin-4-ylcarbonyl)thiophen-2-amine

  • MDL number MFCD03946070
  • Cas Number : 590357-48-5
  • Molecular Weight : 212.2688

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3002607

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3002607
Availability details for
Quantity
  • Availability
  • Add to Cart