CH4238556495 ChemBridge Corporation

2-chloro-1-fluoro-4-isocyanatobenzene

  • MDL number MFCD00037037
  • Cas Number : 50529-33-4
  • Molecular Weight : 171.5562

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3002706

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3002706
Availability details for
Quantity
  • Availability
  • Add to Cart