CH4238556563 ChemBridge Corporation

5,7-dimethyl-1H-indole-2,3-dione

  • MDL number MFCD00047219
  • Cas Number : 39603-24-2
  • Molecular Weight : 175.184

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3002803

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3002803
Availability details for
Quantity
  • Availability
  • Add to Cart