CH4238556602 ChemBridge Corporation

1-(2-fluorobenzyl)-5-methyl-1H-pyrazol-3-amine

  • MDL number MFCD04969964
  • Cas Number : 925146-07-2
  • Molecular Weight : 205.2315

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3002870

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3002870
Availability details for
Quantity
  • Availability
  • Add to Cart