CH4238556635 ChemBridge Corporation

5-cyclopropyl-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-2-carboxylic acid

  • MDL number MFCD03044652
  • Cas Number : 436088-48-1
  • Molecular Weight : 271.1953

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3002919

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3002919
Availability details for
Quantity
  • Availability
  • Add to Cart