CH4238556710 ChemBridge Corporation

(9aS)-octahydro-1H-pyrrolo[1,2-a][1,4]diazepine

  • MDL number MFCD12827460
  • Cas Number : 1240360-64-8
  • Molecular Weight : 140.226

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4017932

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4017932
Availability details for
Quantity
  • Availability
  • Add to Cart