CH4238556850 ChemBridge Corporation

4-(5-methyl-1H-tetrazol-1-yl)benzoic acid

  • MDL number MFCD00458484
  • Cas Number : 64170-57-6
  • Molecular Weight : 204.1854

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/3005046

Safety Information

MSDS https://www.hit2lead.com/building-blocks/3005046
Availability details for
Quantity
  • Availability
  • Add to Cart